Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM16
Entrez Gene ID
102185685
Gene Name
tripartite motif containing 16
Gene Aliases
-
Chromosome Location
Chr.19: 32710749 - 32726721 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693592.3 XP_005693649.2 4 - 5 1218 70
GenBank mRNA KA351406.1 - 4 - 5 1027 70

View Full Product Details