Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCM2
Entrez Gene ID
102188817
Gene Name
glial cells missing transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.23: 5013761 - 5020147 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038751.1 XP_017894240.1 3 - 4 459 78

View Full Product Details