Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASPHD1
Entrez Gene ID
102190009
Gene Name
aspartate beta-hydroxylase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.25: 26266708 - 26271022 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697685.3 XP_005697742.1 2 - 3 1684 64
GenBank mRNA JO634987.2 - 2 - 3 1686 64

View Full Product Details