Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FMR1NB
Entrez Gene ID
102188836
Gene Name
FMR1 neighbor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189596: 305457 - 359779 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700467.2 XP_005700524.2 5 - 6 871 79

View Full Product Details