Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPP25
Entrez Gene ID
102191465
Gene Name
ribonuclease P and MRP subunit p25
Gene Aliases
-
Chromosome Location
Chr.21: 33204225 - 33205677 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005709564.3 XP_005709621.3 1 - 1 745 70

View Full Product Details