Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM82
Entrez Gene ID
102172923
Gene Name
transmembrane protein 82
Gene Aliases
-
Chromosome Location
Chr.16: 50304714 - 50309172 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO397391.2 - 4 - 5 1206 90

View Full Product Details