Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM82
Entrez Gene ID
102172923
Gene Name
transmembrane protein 82
Gene Aliases
-
Chromosome Location
Chr.16: 50304714 - 50309172 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013970310.2 XP_013825764.2 3 - 4 876 81
GenBank mRNA JO397391.2 - 3 - 4 1029 81

View Full Product Details