Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AP1S3
Entrez Gene ID
102184293
Gene Name
adaptor related protein complex 1 subunit sigma 3
Gene Aliases
-
Chromosome Location
Chr.2: 23795517 - 23864914 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058906.1 XP_017914395.1 3 - 4 488 114
XM_018058907.1 XP_017914396.1 3 - 4 493 114

View Full Product Details