Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLX1A
Entrez Gene ID
102184864
Gene Name
SLX1 homolog A, structure-specific endonuclease subunit
Gene Aliases
-
Chromosome Location
Chr.25: 26037255 - 26040604 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO581588.1 - 3 - 4 658 83
JO581589.1 - 3 - 4 658 83
JO581591.1 - 3 - 4 658 83

View Full Product Details