Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102189936
Entrez Gene ID
102189936
Gene Name
Fanconi anemia group B protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017214303: 17786 - 24426 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045530.1 XP_017901019.1 2 - 3 292 74

View Full Product Details