Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LUM
Entrez Gene ID
102176393
Gene Name
lumican
Gene Aliases
-
Chromosome Location
Chr.5: 20785560 - 20795161 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679812.3 XP_005679869.1 2 - 3 1219 94
XM_018047618.1 XP_017903107.1 2 - 3 998 94

View Full Product Details