Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK3A
Entrez Gene ID
102177602
Gene Name
uroplakin 3A
Gene Aliases
-
Chromosome Location
Chr.5: 114611230 - 114619821 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681228.2 XP_005681285.2 1 - 2 93 64

View Full Product Details