Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMUB2
Entrez Gene ID
102180697
Gene Name
transmembrane and ubiquitin like domain containing 2
Gene Aliases
-
Chromosome Location
Chr.19: 43642345 - 43646310 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693919.3 XP_005693976.1 1 - 2 600 74

View Full Product Details