Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WNT10B
Entrez Gene ID
102169259
Gene Name
Wnt family member 10B
Gene Aliases
-
Chromosome Location
Chr.5: 30483587 - 30489705 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047850.1 XP_017903339.1 2 - 3 1101 66

View Full Product Details