Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102181933
Entrez Gene ID
102181933
Gene Name
ovochymase-1
Gene Aliases
OVCH1
Chromosome Location
Chr.5: 78663813 - 78731318 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049053.1 XP_017904542.1 7 - 8 855 96

View Full Product Details