Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VCPIP1
Entrez Gene ID
102168647
Gene Name
valosin containing protein interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.14: 51223503 - 51252791 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013969192.2 XP_013824646.1 1 - 2 2981 94

View Full Product Details