Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTDSP1
Entrez Gene ID
102176569
Gene Name
CTD small phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.2: 29390623 - 29397856 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059502.1 XP_017914991.1 5 - 6 1818 82
XM_018059509.1 XP_017914998.1 5 - 6 1818 82
GenBank mRNA JO637725.2 - 5 - 6 710 82

View Full Product Details