Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRSS27
Entrez Gene ID
102174527
Gene Name
serine protease 27
Gene Aliases
-
Chromosome Location
Chr.25: 2017033 - 2029105 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040362.1 XP_017895851.1 1 - 2 152 122
GenBank mRNA KA339530.1 - 1 - 2 222 122

View Full Product Details