Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDXACB1
Entrez Gene ID
102170993
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 60144704 - 60149555 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689433.3 XP_005689490.2 4 - 5 721 92

View Full Product Details