Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD79A
Entrez Gene ID
102174687
Gene Name
CD79a molecule
Gene Aliases
-
Chromosome Location
Chr.18: 52244524 - 52248386 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692572.3 XP_005692629.1 4 - 5 799 92
GenBank mRNA JO341774.2 - 1 - 2 461 92

View Full Product Details