Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM81A
Entrez Gene ID
102178453
Gene Name
family with sequence similarity 81 member A
Gene Aliases
-
Chromosome Location
Chr.10: 52255870 - 52335510 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054284.1 XP_017909773.1 5 - 6 555 103
XM_018054285.1 XP_017909774.1 4 - 5 549 103
XM_018054286.1 XP_017909775.1 5 - 6 564 103
XM_018054287.1 XP_017909776.1 4 - 5 595 103
GenBank mRNA JO436387.2 - 5 - 6 833 103

View Full Product Details