Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MZT1
Entrez Gene ID
102178637
Gene Name
mitotic spindle organizing protein 1
Gene Aliases
-
Chromosome Location
Chr.12: 38934235 - 38948901 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687650.3 XP_005687707.1 2 - 3 339 86
GenBank mRNA JO638058.2 - 2 - 3 319 86

View Full Product Details