Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD6
Entrez Gene ID
102180538
Gene Name
caspase recruitment domain family member 6
Gene Aliases
-
Chromosome Location
Chr.20: 33613839 - 33627750 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694755.2 XP_005694812.2 2 - 3 850 83
GenBank mRNA JT672744.1 - 4 - 5 2505 83

View Full Product Details