Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDKN2AIP
Entrez Gene ID
102187189
Gene Name
CDKN2A interacting protein
Gene Aliases
-
Chromosome Location
Chr.27: 31278808 - 31282373 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698752.3 XP_005698809.3 2 - 3 580 106

View Full Product Details