Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTSO
Entrez Gene ID
102168847
Gene Name
cathepsin O
Gene Aliases
-
Chromosome Location
Chr.17: 27751013 - 27796181 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061204.1 XP_017916693.1 3 - 4 383 83
GenBank mRNA KA351649.1 - 6 - 7 1357 83

View Full Product Details