Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEG3
Entrez Gene ID
102189790
Gene Name
paternally expressed 3
Gene Aliases
-
Chromosome Location
Chr.18: 65561895 - 65584442 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063405.1 XP_017918894.1 6 - 7 2265 89
XM_018063406.1 XP_017918895.1 6 - 7 2265 89
XM_018063407.1 XP_017918896.1 7 - 8 1084 89
XM_018063408.1 XP_017918897.1 6 - 7 1021 89
XM_018063409.1 XP_017918898.1 6 - 7 711 89
GenBank mRNA JO426493.1 - 6 - 7 795 89

View Full Product Details