Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT9B
Entrez Gene ID
102169419
Gene Name
Wnt family member 9B
Gene Aliases
-
Chromosome Location
Chr.19: 44964194 - 44986625 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064015.1 XP_017919504.1 7 - 8 1210 57

View Full Product Details