Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS5
Entrez Gene ID
102172793
Gene Name
integrator complex subunit 5
Gene Aliases
-
Chromosome Location
Chr.29: 41313918 - 41318567 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013975915.2 XP_013831369.1 1 - 2 145 79
GenBank mRNA JO406288.2 - 1 - 2 795 79
JT668623.1 - 1 - 2 3110 79

View Full Product Details