Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASL10A
Entrez Gene ID
102190251
Gene Name
RAS like family 10 member A
Gene Aliases
-
Chromosome Location
Chr.17: 2666273 - 2669244 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691714.3 XP_005691771.2 2 - 3 835 89

View Full Product Details