Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C8H9orf24
Entrez Gene ID
102184719
Gene Name
chromosome 8 C9orf24 homolog
Gene Aliases
-
Chromosome Location
Chr.8: 75524267 - 75539046 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052386.1 XP_017907875.1 3 - 4 785 88

View Full Product Details