Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCK1
Entrez Gene ID
102189995
Gene Name
NCK adaptor protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 131805588 - 131884444 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050814.1 XP_017906303.1 2 - 3 367 81
XM_018050821.1 XP_017906310.1 2 - 3 370 81
XM_018050830.1 XP_017906319.1 2 - 3 341 81
XM_018050839.1 XP_017906328.1 2 - 3 350 81
GenBank mRNA JO610951.2 - 2 - 3 1324 81

View Full Product Details