Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OGFRL1
Entrez Gene ID
102184825
Gene Name
opioid growth factor receptor like 1
Gene Aliases
-
Chromosome Location
Chr.14: 92065054 - 92084741 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058828.1 XP_017914317.1 4 - 5 573 68
XM_018058829.1 XP_017914318.1 4 - 5 624 68
GenBank mRNA JO631973.2 - 4 - 5 679 68

View Full Product Details