The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018045144.1 | XP_017900633.1 | - | 672 | 84 |
XM_018045145.1 | XP_017900634.1 | - | 672 | 84 |