The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005677143.3 | XP_005677200.2 | 3 - 4 | 305 | 58 |
GenBank mRNA | JO612023.2 | - | 4 - 5 | 1834 | 58 |