Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11H2orf42
Entrez Gene ID
102189409
Gene Name
chromosome 11 C2orf42 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 68201711 - 68246551 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686840.2 XP_005686897.1 9 - 10 1864 89
XM_005686841.3 XP_005686898.1 10 - 11 4526 89
XM_013967789.2 XP_013823243.1 9 - 10 1885 89
GenBank mRNA JT701341.1 - 5 - 6 912 89

View Full Product Details