Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POU2AF1
Entrez Gene ID
102169494
Gene Name
POU class 2 associating factor 1
Gene Aliases
-
Chromosome Location
Chr.15: 60709078 - 60734672 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059807.1 XP_017915296.1 2 - 3 338 100

View Full Product Details