Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRP15
Entrez Gene ID
102175604
Gene Name
ribosomal RNA processing 15 homolog
Gene Aliases
-
Chromosome Location
Chr.16: 20279460 - 20332663 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690476.3 XP_005690533.3 2 - 3 457 99
GenBank mRNA JO585333.2 - 3 - 4 550 99
KA309982.1 - 3 - 4 619 99

View Full Product Details