Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17H5orf52
Entrez Gene ID
102191452
Gene Name
chromosome 17 C5orf52 homolog
Gene Aliases
-
Chromosome Location
Chr.17: 1682600 - 1687157 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691637.3 XP_005691694.3 1 - 2 341 119

View Full Product Details