Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLDN9
Entrez Gene ID
102188006
Gene Name
claudin 9
Gene Aliases
-
Chromosome Location
Chr.25: 2312129 - 2313224 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697412.2 XP_005697469.1 1 - 1 641 65

View Full Product Details