Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IDO2
Entrez Gene ID
102184590
Gene Name
indoleamine 2,3-dioxygenase 2
Gene Aliases
-
Chromosome Location
Chr.27: 10480761 - 10549730 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698880.3 XP_005698937.1 1 - 2 228 122
GenBank mRNA JO397671.2 - 9 - 10 1463 122
JO432418.2 - 8 - 9 1507 122

View Full Product Details