Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IDO2
Entrez Gene ID
102184590
Gene Name
indoleamine 2,3-dioxygenase 2
Gene Aliases
-
Chromosome Location
Chr.27: 10480761 - 10549730 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698880.3 XP_005698937.1 8 - 9 848 97
XM_005698881.3 XP_005698938.1 7 - 8 752 97
XM_018042134.1 XP_017897623.1 7 - 8 1180 97
GenBank mRNA JO397671.2 - 2 - 3 843 97
JO432418.2 - 2 - 3 887 97

View Full Product Details