Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEDD2
Entrez Gene ID
102173878
Gene Name
death effector domain containing 2
Gene Aliases
-
Chromosome Location
Chr.18: 51942729 - 51958186 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062497.1 XP_017917986.1 3 - 4 619 72
GenBank mRNA KA332933.1 - 2 - 3 1347 72

View Full Product Details