Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYG1
Entrez Gene ID
102184731
Gene Name
lysozyme g1
Gene Aliases
-
Chromosome Location
Chr.11: 4315996 - 4325224 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686285.2 XP_005686342.1 1 - 2 40 58

View Full Product Details