Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCLAT1
Entrez Gene ID
102191623
Gene Name
lysocardiolipin acyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.11: 68969224 - 69159749 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055477.1 XP_017910966.1 5 - 6 775 90
XM_018055478.1 XP_017910967.1 4 - 5 606 90
GenBank mRNA JT762624.1 - 4 - 5 524 90

View Full Product Details