Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ID2
Entrez Gene ID
102189873
Gene Name
inhibitor of DNA binding 2
Gene Aliases
-
Chromosome Location
Chr.11: 88211639 - 88214135 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687031.3 XP_005687088.1 2 - 3 626 62
GenBank mRNA JO579891.1 - - 970 62

View Full Product Details