Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZIM3
Entrez Gene ID
102190350
Gene Name
zinc finger imprinted 3
Gene Aliases
-
Chromosome Location
Chr.18: 65840960 - 65847359 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063254.1 XP_017918743.1 2 - 3 185 70

View Full Product Details