Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCF2
Entrez Gene ID
102188716
Gene Name
IQ motif containing F2
Gene Aliases
-
Chromosome Location
Chr.22: 49112781 - 49114290 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695901.1 XP_005695958.1 2 - 3 135 83

View Full Product Details