Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2G12A
Entrez Gene ID
102184325
Gene Name
phospholipase A2 group XIIA
Gene Aliases
-
Chromosome Location
Chr.6: 15978599 - 15992994 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013964470.2 XP_013819924.1 2 - 3 455 67
XM_018049225.1 XP_017904714.1 2 - 3 466 67

View Full Product Details