Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
C18H19orf47
Entrez Gene ID
102186989
Gene Name
chromosome 18 C19orf47 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 50585683 - 50606607 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062416.1 XP_017917905.1 5 - 6 451 65
XM_018062417.1 XP_017917906.1 5 - 6 435 65
XM_018062418.1 XP_017917907.1 5 - 6 451 65
XM_018062419.1 XP_017917908.1 5 - 6 451 65

View Full Product Details