Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZIM2
Entrez Gene ID
102189239
Gene Name
zinc finger imprinted 2
Gene Aliases
-
Chromosome Location
Chr.18: 65471223 - 65486285 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063252.1 XP_017918741.1 5 - 6 822 75

View Full Product Details